Folpet Analytical standard, reference material, Fluka™
Catalog # HONCR1891-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:133-07-3
- Popis:Folpet
- Podmínky uskladnění:RT
- MDL čí.:MFCD00047303
- Synonym(s):2-[(Trichloromethyl)thio]-1H-isoindole-1,3(2H)-dione
- Vzorec:C₉H₄Cl₃NO₂S
- Molekulární hmotnost:296.56 g/mol
- Popis:Folpet
- Fyzická forma:Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery