Terbuthylazine Analytical standard, reference material, Fluka™
HONCR1885-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:5915-41-3
- Popis:Terbuthylazine
- Podmínky uskladnění:RT
- MDL čí.:MFCD00055513
- Synonym(s):6-Chloro-N2-(1,1-dimethylethyl)-N4-ethyl-1,3,5-triazine-2,4-diamine
- Bod tání:175 °C
- Vzorec:C₉H₁₆ClN₅
- Molekulární hmotnost:229.71 g/mol
- Popis:Terbuthylazine
- Fyzická forma:Solid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery