Dexamethasone Analytical standard, reference material, Fluka™
Catalog # HONCR1879-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:50-02-2
- Popis:Dexamethasone
- Podmínky uskladnění:RT
- MDL čí.:MFCD00064136
- Synonym(s):(11ß,16a)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione
- Vzorec:C₂₂H₂₉FO₅
- Popis:Dexamethasone
- Fyzická forma:Crystalline
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery