Trimethoprim Analytical standard, reference material, Fluka™
HONCR1873-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:738-70-5
- Popis:Trimethoprim
- Podmínky uskladnění:2-8°C
- MDL čí.:MFCD00036761
- Synonym(s):2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine
- Bod tání:200 °C
- Vzorec:C₁₄H₁₈N₄O₃
- Molekulární hmotnost:290.32 g/mol
- Popis:Trimethoprim
- Fyzická forma:Solid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery