Enrofloxacin Analytical standard, reference material, Fluka™
HONCR1871-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:93106-60-6
- Popis:Enrofloxacin
- Podmínky uskladnění:RT
- MDL čí.:MFCD00792463
- Synonym(s):1,4-Dihydro-1-cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-4-oxo-3-quinolinecarboxylic acid
- Bod tání:223 °C
- Vzorec:C₁₉H₂₂FN₃O₃
- Molekulární hmotnost:359.39 g/mol
- Popis:Enrofloxacin
- Fyzická forma:Solid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery