Oxytetracycline dihydrate Analytical standard, reference material, Fluka™
Catalog # HONCR1869-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:6153-64-6
- Popis:Oxytetracycline dihydrate
- Podmínky uskladnění:RT
- MDL čí.:MFCD00151234
- Synonym(s):4-(Dimethylamino)-1,4,4a,5,-5a,6,11,12a-octahydro-3,5,6,-10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-2-naphtacencarboxamid
- Vzorec:C₂₂H₂₄N₂O₉
- Popis:Oxytetracycline dihydrate
- Fyzická forma:Crystalline
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery