Phorate sulphoxide Analytical standard, reference material, Fluka™
Catalog # HONCR1866-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:50 mg
- Standard:Analytical standard
- Čí. CAS:2588-03-6
- Popis:Phorate sulphoxide
- Podmínky uskladnění:2-8°C
- MDL čí.:MFCD01705308
- Č. UN:2810
- Synonym(s):O,O-Diethyl S-eththionylmethyl thiothionophosphate
- Vzorec:C₇H₁₇O₃PS₃
- Molekulární hmotnost:276.38 g/mol
- Popis:Phorate sulphoxide
- Fyzická forma:Viscous Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery