Phorate-oxon sulphone Analytical standard, reference material, Fluka™
HONCR1864-25MG
:
- Typ balení:Glass Bottle
- Bal.:25 mg
- Standard:Analytical standard
- Čí. CAS:2588-06-9
- Popis:Phorate-oxon sulphone
- Podmínky uskladnění:2-8°C
- Č. UN:2810
- Synonym(s):O,O-Diethyl S-(ethylsulfonyl)methyl phosphorothioate; Phosphorothioic Acid O,O-Diethyl S-[(Ethylsulfonyl)methyl] Ester
- Vzorec:C₇H₁₇O₅PS₂
- Molekulární hmotnost:276.31 g/mol
- Popis:Phorate-oxon sulphone
- Fyzická forma:Viscous Liquid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery