Pencycuron Analytical standard, reference material, Fluka™
Catalog # HONCR1863-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:66063-05-6
- Popis:Pencycuron
- Podmínky uskladnění:RT
- MDL čí.:MFCD00078724
- Synonym(s):1-(4-chlorobenzyl)-1-cyclopentyl-3-phenylurea
- Bod tání:132 °C
- Vzorec:C₁₉H₂₁ClN₂O
- Molekulární hmotnost:328.84 g/mol
- Popis:Pencycuron
- Bod vzplanutí:not auto-flammable
- Fyzická forma:Crystalline
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery