Cyhalofop-butyl Analytical standard, reference material, Fluka™
Catalog # HONCR1862-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:50 mg
- Standard:Analytical standard
- Čí. CAS:122008-85-9
- Popis:Cyhalofop-butyl
- Podmínky uskladnění:RT
- MDL čí.:MFCD01631151
- Synonym(s):Butyl (R)-2-[4-(4-cyano-2-fluorophenoxy)phenoxy]propionate
- Bod tání:49.5 °C
- Vzorec:C₂₀H₂₀FNO₄
- Molekulární hmotnost:357.38 g/mol
- Popis:Cyhalofop-butyl
- Bod vzplanutí:122 °C.closed cup
- Fyzická forma:Crystalline
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery