Isotianil Analytical standard, reference material, Fluka™
Catalog # HONCR1861-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:224049-04-1
- Popis:Isotianil
- Podmínky uskladnění:RT
- MDL čí.:MFCD16652537
- Synonym(s):3,4-Dichloro-2'-cyano-1,2-thiazole-5-carboxanilide
- Bod tání:196 °C
- Vzorec:C₁₁H₅Cl₂N₃OS
- Molekulární hmotnost:298.15 g/mol
- Popis:Isotianil
- Fyzická forma:powder
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery