Pyridaben Analytical standard, reference material, Fluka™
HONCR1859-25MG
:
- Typ balení:Glass Bottle
- Bal.:25 mg
- Standard:Analytical standard
- Čí. CAS:96489-71-3
- Popis:Pyridaben
- Podmínky uskladnění:RT
- MDL čí.:MFCD00274601
- Č. UN:2811
- Synonym(s):4-chloro-2-(1,1-dimethylethyl)-5-[[[4-(1,1-dimethylethyl)phenyl]methyl]thio]-3(2H)-pyridazinone
- Bod tání:111 - 112 °C
- Vzorec:C₁₉H₂₅ClN₂OS
- Molekulární hmotnost:364.93 g/mol
- Popis:Pyridaben
- Fyzická forma:Crystalline
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery