Phosmet Analytical standard, reference material, Fluka™
Katalogové číslo HONCR1854-100MG
Dodavatel: Honeywell Chemicals
Číslo CAS:
Specifikace
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:732-11-6
- Popis:Phosmet
- Podmínky uskladnění:2-8°C
- MDL čí.:MFCD00055501
- Č. UN:2811
- Synonym(s):O,O-Dimethyl S-(phthalimidomethyl) phosphorodithioate
- Bod tání:71.8 °C
- Vzorec:C₁₁H₁₂NO₄PS₂
- Molekulární hmotnost:317.32 g/mol
- Popis:Phosmet
- Fyzická forma:Solid
Specifikace
O tomto produktu
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery