Chlorfenvinphos (Isomerengemisch Z+E) Analytical standard, reference material, Fluka™
HONCR1850-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:470-90-6
- Popis:Chlorfenvinphos (Isomerengemisch Z+E)
- Podmínky uskladnění:2-8°C
- MDL čí.:MFCD00041818
- Č. UN:2810
- Synonym(s):(EZ)-2-chloro-1-(2,4-dichlorophenyl)vinyl diethyl phosphate
- Bod tání:-23 - -19 °C
- Vzorec:C₁₂H₁₄Cl₃O₄P
- Molekulární hmotnost:359.57 g/mol
- Popis:Chlorfenvinphos (Isomerengemisch Z+E)
- Bod vzplanutí:> 285 °C
- Fyzická forma:liquid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery