Spiromesifen Analytical standard, reference material, Fluka™
Catalog # HONCR1849-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:283594-90-1
- Popis:Spiromesifen
- Podmínky uskladnění:RT
- MDL čí.:MFCD08458959
- Synonym(s):3-Mesityl-2-oxo-1-oxaspiro[4.4]non-3-en-4-yl) 3,3-dimethylbutyrate
- Bod tání:96.7 - 98.7 °C
- Vzorec:C₂₃H₃₀O₄
- Molekulární hmotnost:370.48 g/mol
- Popis:Spiromesifen
- Fyzická forma:Crystalline
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery