Penflufen Analytical standard, reference material, Fluka™
Katalogové číslo HONCR1847-100MG
Dodavatel: Honeywell Chemicals
Číslo CAS:
Specifikace
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:494793-67-8
- Popis:Penflufen
- Podmínky uskladnění:RT
- MDL čí.:MFCD22200854
- Synonym(s):2'-[(RS)-1,3-Dimethylbutyl]-5-fluoro-1,3-dimethylpyrazole-4-carboxanilide
- Bod tání:111 °C
- Vzorec:C₁₈H₂₄FN₃O
- Molekulární hmotnost:317.40 g/mol
- Popis:Penflufen
- Bod vzplanutí:100 °C
- Fyzická forma:Crystalline
Specifikace
O tomto produktu
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery