Spirotetramat Analytical standard, reference material, Fluka™
Catalog # HONCR1845-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:203313-25-1
- Popis:Spirotetramat
- Podmínky uskladnění:RT
- Synonym(s):cis-3-(2,5-Dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro[4.5]dec-3-en-4-yl ethyl carbonate
- Bod tání:142 °C
- Vzorec:C₂₁H₂₇NO₅
- Molekulární hmotnost:373.44 g/mol
- Popis:Spirotetramat
- Fyzická forma:Powder
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery