Order Entry
Czech Republic
ContactUsLinkComponent
Flubendiamide Analytical standard, reference material, Fluka™
Flubendiamide Analytical standard, reference material, Fluka™
Catalog # HONCR1842-100MG
CAS Number:  
Flubendiamide Analytical standard, reference material, Fluka™
Catalog # HONCR1842-100MG
CAS Number:  

Specifications

  • Typ balení:
    Glass Bottle
  • Bal.:
    100 mg
  • Standard:
    Analytical standard
  • Čí. CAS:
    272451-65-7
  • Popis:
    Flubendiamide
  • Podmínky uskladnění:
    RT
  • MDL čí.:
    MFCD09953911
  • Synonym(s):
    N2-[1,1-Dimethyl-2-(methylsulfonyl)ethyl]-3-iodo-N1-{2-methyl-4-[1,2,2,2-tetrafluoro-1-(trifluoromethyl)ethyl]phenyl}-1,2-benzenedicarboxamide
  • Bod tání:
    217 - 221 °C
  • Vzorec:
    C₂₃H₂₂F₇IN₂O₄S
  • Molekulární hmotnost:
    682.39 g/mol
  • Popis:
    Flubendiamide
  • Fyzická forma:
    Powder

Specifications

About this item

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery