Penconazole Analytical standard, reference material, Fluka™
HONCR1841-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:66246-88-6
- Popis:Penconazole
- Podmínky uskladnění:RT
- MDL čí.:MFCD00078737
- Synonym(s):1-[2-(2,4-dichlorophenyl)pentyl]-1H-1,2,4-triazole
- Bod tání:60.3 - 61 °C
- Vzorec:C₁₃H₁₅Cl₂N₃
- Molekulární hmotnost:284.2 g/mol
- Popis:Penconazole
- Fyzická forma:Powder
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery