Simazine Analytical standard, reference material, Fluka™
HONCR1833-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:122-34-9
- Popis:Simazine
- Podmínky uskladnění:RT
- MDL čí.:MFCD00023174
- Synonym(s):2,4-Bis(ethylamino)-6-chloro-1,3,5-triazine
- Bod tání:225 - 227 °C
- Vzorec:C₇H₁₂ClN₅
- Molekulární hmotnost:201.66 g/mol
- Popis:Simazine
- Fyzická forma:Solid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery