Order Entry
Czech Republic
ContactUsLinkComponent
Mecoprop Analytical standard, reference material, Fluka™
Mecoprop Analytical standard, reference material, Fluka™
Catalog # HONCR1831-100MG
CAS Number:  
Mecoprop Analytical standard, reference material, Fluka™
Catalog # HONCR1831-100MG
CAS Number:  

Specifications

  • Typ balení:
    Glass Bottle
  • Bal.:
    100 mg
  • Standard:
    Analytical standard
  • Čí. CAS:
    93-65-2
  • Popis:
    Mecoprop
  • Podmínky uskladnění:
    RT
  • MDL čí.:
    MFCD00002648
  • Synonym(s):
    2-(4-Chloro-2-methylphenoxy)propionic acid
  • Vzorec:
    C₁₀H₁₁ClO₃
  • Popis:
    Mecoprop
  • Fyzická forma:
    Crystalline

Specifications

About this item

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery