Prosulphocarb Analytical standard, reference material, Fluka™
Catalog # HONCR1830-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:52888-80-9
- Popis:Prosulphocarb
- Podmínky uskladnění:RT
- MDL čí.:MFCD00145179
- Synonym(s):S-benzyl dipropyl(thiocarbamate)
- Bod tání:-20 °C
- Vzorec:C₁₄H₂₁NOS
- Molekulární hmotnost:251.39 g/mol
- Popis:Prosulphocarb
- Bod vzplanutí:132 °C
- Fyzická forma:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery