Microcystin-LY- solution, 6-8 μg/mL in methanol Analytical standard, reference material, Fluka™
Catalog # HONCR1827-1ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:1 mL
- Standard:Analytical standard
- Čí. CAS:67-56-1
- Popis:Microcystin-LY- solution, 6-8 μg/mL in methanol
- Podmínky uskladnění:Ambient
- MDL čí.:MFCD10567358
- Č. UN:1230
- Synonym(s):15-[(4-hydroxyphenyl)methyl]-18-[(1Z,3E)-6-methoxy-3,5-dimethyl-7-phenylhepta-1,3-dienyl]-1,5,12,19-tetramethyl-2-methylidene-8-(2-methylpropyl)-3,6,9,13,16,20,25-heptaoxo-1,4,7,10,14,17,21-heptazacyclopentacosane-11,22-dicarboxylic acid; Algae bloom toxin, Biotoxin, Cyanobacterial toxin
- Bod tání:-98 °C
- Vzorec:C₅₂H₇₁N₇O₁₃
- Koncentrace:6-8 μg/mL in methanol
- Popis:Microcystin-LY
- Bod vzplanutí:11 °C
- Fyzická forma:liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery