Microcystin-LR-solution, 10 μg/mL in methanol Analytical standard, reference material, Fluka™
Catalog # HONCR1823-1ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:1 mL
- Standard:Analytical standard
- Čí. CAS:101043-37-2
- Popis:Microcystin-LR-solution, 10 μg/mL in methanol
- Podmínky uskladnění:Ambient
- MDL čí.:MFCD01774058
- Č. UN:1230
- Synonym(s):(5R,8S,11R,12S,15S,18S,19S,22R)-15-[3-(diaminomethylideneamino)propyl]-18-[(1E,3E,5S,6S)-6-methoxy-3,5-dimethyl-7-phenylhepta-1,3-dienyl]-1,5,12,19-tetramethyl-2-methylidene-8-(2-methylpropyl)-3,6,9,13,16,20,25-heptaoxo-1,4,7,10,14,17,21-heptazacyclopentacosane-11,22-dicarboxylic acid
- Bod tání:-98 °C
- Vzorec:C₄₉H₇₄N₁₀O₁₂
- Koncentrace:10 μg/mL in methanol
- Popis:Microcystin-LR
- Bod vzplanutí:11 °C
- Fyzická forma:liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery