Phorate-oxon Analytical standard, reference material, Fluka™
Catalog # HONCR1817-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:25 mg
- Standard:Analytical standard
- Čí. CAS:2600-69-3
- Popis:Phorate-oxon
- Podmínky uskladnění:2-8°C
- Č. UN:2810
- Synonym(s):O,O-Diethyl S-(ethylthiomethyl) phosphorothioate
- Vzorec:C₇H₁₇O₃PS₂
- Molekulární hmotnost:244.31 g/mol
- Popis:Phorate-oxon
- Fyzická forma:Viscous Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery