Temephos Analytical standard, reference material, Fluka™
HONCR1816-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:3383-96-8
- Popis:Temephos
- Podmínky uskladnění:2-8°C
- MDL čí.:MFCD00128034
- Synonym(s):O,O,O',O'-tetramethyl O,O'-thiodi-p-phenylene bis(phosphorothioate)
- Vzorec:C₁₆H₂₀O₆P₂S₃
- Molekulární hmotnost:466.47 g/mol
- Popis:Temephos
- Fyzická forma:Crystalline
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery