Phorate Analytical standard, reference material, Fluka™
HONCR1808-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:298-02-2
- Popis:Phorate
- Podmínky uskladnění:2-8°C
- MDL čí.:MFCD00036348
- Č. UN:2810
- Synonym(s):O,O-Diethyl S-(ethylthiomethyl) phosphorodithioate
- Bod tání:-15 °C
- Vzorec:C₇H₁₇O₂PS₃
- Molekulární hmotnost:260.4 g/mol
- Popis:Phorate
- Bod vzplanutí:closed cup.110 °C
- Fyzická forma:Liquid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery