Fluazifop-P-butyl Analytical standard, reference material, Fluka™
HONCR1802-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:79241-46-6
- Popis:Fluazifop-P-butyl
- Podmínky uskladnění:RT
- MDL čí.:MFCD06199153
- Bod tání:-46 °C
- Molekulární hmotnost:383.36 g/mol
- Popis:Fluazifop-P-butyl
- Fyzická forma:Liquid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery