Bifenthrin Analytical standard, reference material, Fluka™
Catalog # HONCR1798-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:82657-04-3
- Popis:Bifenthrin
- Podmínky uskladnění:RT
- Č. UN:2811
- Bod tání:66.6 - 69 °C
- Molekulární hmotnost:422.87 g/mol
- Popis:Bifenthrin
- Fyzická forma:Semi-Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery