Fluxapyroxad Analytical standard, reference material, Fluka™
HONCR1797-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:907204-31-3
- Popis:Fluxapyroxad
- Podmínky uskladnění:RT
- MDL čí.:MFCD22373628
- Bod tání:157 °C
- Molekulární hmotnost:381.30 g/mol
- Popis:Fluxapyroxad
- Fyzická forma:Powder
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery