GemCell Plus™ U.S. Human Serum AB - Xeno-Free
Katalogové číslo 100-912-100
Dodavatel: GEMINI BIO-PRODUCTS
Specifikace
- Popis:U.S. human serum AB
- Sekvence:01
- Původ:US
- Skladovací teplota:Store frozen at −20…−10 °C
- Typ balení:Bottle
- Bal.:100 mL
Specifikace
O tomto produktu
GemCell Plus™ Xeno-Free Human Serum is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material goes through additional rigorous testing and contains therapeutic grade human thrombin. All donor units are tested for viral markers and found to be non-reactive.
- Developed without the use of animal material
- Gamma irradiation available