ContactUsLinkComponent
Carvedilol Related Compounds, Reference Standard
Carvedilol Related Compounds, Reference Standard
Supplier:  USP
Certificates
Carvedilol Related Compounds, Reference Standard
Supplier:  USP
Specifications
  • Čí. CAS:
    2057412-43-6
  • Synonym(s):
    2-(2-Methoxyphenoxy)ethyl amine hydrochloride monohydrate
  • Vzorec:
    C₉H₁₃NO₂
  • Popis:
    Carvedilol Related Compound E