Orders LinkContactUsLinkComponent
Sodium Acetate, Reference Standards
Sodium Acetate, Reference Standards
Dodavatel:   USP
Sodium Acetate, Reference Standards
Dodavatel:  USP
Certifikáty
2 Dostupných variant níže
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Documents
Specifications for Product Family
Specifikace
Specifikace
  • Catalog No:
  • 1613407.
  • 1613429.
  • Popis:
  • Sodium Acetate
  • Sodium Acetate Trihydrate
  • Čí. CAS:
  • 127-09-3
  • 6131-90-4
Specifikace
Product Family Options
Informace o produktu
  • Standard
    Typ balení
    Bal.
    Dostupnost
    Price
  • Sodium Acetate
    USP reference standard
    Vial
    1 g
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 g
    1 * 1 g - 8 840,00 Kč
    8 840,00 Kč
    1 * 1 g
       8 840,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1613407.
    Popis
    Sodium Acetate
    Čí. CAS
    127-09-3
  • 1613429.
    Sodium Acetate Trihydrate
    Sodium Acetate Trihydrate
    USP reference standard
    Vial
    1 g
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 g
    1 * 1 g - 13 830,00 Kč
    13 830,00 Kč
    1 * 1 g
       13 830,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    1613429.
    Popis
    Sodium Acetate Trihydrate
    Čí. CAS
    6131-90-4