Orders LinkContactUsLinkComponent
Eleuthero Extract, Reference Standard
Eleuthero Extract, Reference Standard
Powdered Eleuthero Extract, USP reference standard, Vial, 1,5 g
Katalogové číslo 1234704.
Dodavatel:   USP
Eleuthero Extract, Reference Standard
Katalogové číslo 1234704.
Dodavatel:  USP
Číslo CAS:  
Eleuthero Extract, Reference Standard
Powdered Eleuthero Extract, USP reference standard, Vial, 1,5 g
Přejít na:
Číslo CAS:  
Price
15 650,00 Kč
Přihlásit se pro dostupnost
1 * 1,5 g
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifikace
Specifikace
  • Typ balení:
    Vial
  • Bal.:
    1,5 g
  • Standard:
    USP reference standard
  • Popis:
    Powdered Eleuthero Extract
  • Popis:
    Eleuthero
  • Cat. No.:
    1234704.
  • Čí. CAS:
    84696-12-5