Insulin, Reference Standards
O tomto produktu
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Insulin Glargine for Peak Identication: Mixture of Insulin Glargine and 0A-Arg-Insulin Glargine
Certifications: US Pharmacopea Reference Standard
Upozornení: Cold Shipment required
6 Dostupných variant níže
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Insulin Glargine for Peak Identication: Mixture of Insulin Glargine and 0A-Arg-Insulin Glargine
Certifications: US Pharmacopea Reference Standard
Upozornení: Cold Shipment required
Documents
Certifikáty
Hledat podle čísla šaržeSpecifications for Product Family
Specifikace
Specifikace
- Catalog No:
- 1342300.
- 1342321.
- 1342037.
- 1342150.
- 1342060.
- 1342059.
- Popis:
- Insulin Pork
- Insulin Lispro
- Insulin Aspart
- High Molecular Weight Insulin Human
- Insulin Glargine for Peak Identication
- Insulin Glargine
- Čí. CAS:
- 12584-58-6
- 133107-64-9
- 116094-23-6
- 11061-68-0
- 160337-95-1
Specifikace
Product Family Options
Informace o produktu
- StandardTyp baleníBal.DostupnostPrice
- 1342300.Insulin PorkSpecifikace:
More
Specifications
Cat. No.1342300.PopisInsulin PorkČí. CAS12584-58-6 - 1342321.Insulin LisproSpecifikace:
More
Specifications
Cat. No.1342321.PopisInsulin LisproČí. CAS133107-64-9 - 1342037.Insulin AspartSpecifikace:
More
Specifications
Cat. No.1342037.PopisInsulin AspartČí. CAS116094-23-6 - 1342150.High Molecular Weight Insulin HumanHigh Molecular Weight Insulin HumanUSP reference standardAmpule8,4 mgPřihlásit se pro dostupnost1 * 8,4 mg27 940,00 KčSpecifikace:
More
Specifications
Cat. No.1342150.PopisHigh Molecular Weight Insulin HumanČí. CAS11061-68-0 - 1342060.Insulin Glargine for Peak IdenticationInsulin Glargine for Peak IdenticationUSP reference standardAmpule3,2 mgPřihlásit se pro dostupnost1 * 3,2 mg30 840,00 KčSpecifikace:
More
Specifications
Cat. No.1342060.PopisInsulin Glargine for Peak IdenticationČí. CAS - 1342059.Insulin GlargineInsulin GlargineUSP reference standardAmpule15,6 mgPřihlásit se pro dostupnost1 * 15,6 mg10 260,00 KčSpecifikace:
More
Specifications
Cat. No.1342059.PopisInsulin GlargineČí. CAS160337-95-1