0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Centrifugy
Rotory
Zonal Rotor with Standard Core, Titanium, Ti 15, Optima
Zonal Rotor with Standard Core, Titanium, Ti 15, Optima
521-4164
:
Beckman Coulter
Zonal Rotor with Standard Core, Titanium, Ti 15, Optima
521-4164
:
Beckman Coulter
Popis:
Ti-15 Zonal rotor with standard core, titanium
Environmentally Preferable:
Bal.:
1
Určeno pro:
Swinging bucket (High Performance)
Max. RPM:
32000
Poloměr:
88,9 mm