0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Centrifugy
Rotory
Swinging-bucket rotor, SW 41 Ti, Optima
Swinging-bucket rotor, SW 41 Ti, Optima
521-3575
:
Beckman Coulter
Swinging-bucket rotor, SW 41 Ti, Optima
521-3575
:
Beckman Coulter
Popis:
SW 41 Ti swinging bucket rotor
Bal.:
1
Určeno pro:
Swinging bucket (ultracentrifugation)
Max. RPM:
41000
Poloměr:
153,1 mm