Orders LinkContactUsLinkComponent
Rotor Package, MLN-80 NVT, Optima
Rotor Package, MLN-80 NVT, Optima
MLN-80 NVT rotor package, 80000 rpm, 389000 ×g, 8×8 ml. Major applications: Rapid contamination-free isopycnic isolation of plasmid DNA. For use in Optima MAX-XP benchtop ultracentrifuges and Optima MAX ultracentrifuges, Near vertical (ultracentrifugation)
Katalogové číslo 521-4042
Dodavatel:   Beckman Coulter
Rotor Package, MLN-80 NVT, Optima
Katalogové číslo 521-4042
Dodavatel:  Beckman Coulter
Rotor Package, MLN-80 NVT, Optima
Rotor Package, MLN-80 NVT, Optima
MLN-80 NVT rotor package, 80000 rpm, 389000 ×g, 8×8 ml. Major applications: Rapid contamination-free isopycnic isolation of plasmid DNA. For use in Optima MAX-XP benchtop ultracentrifuges and Optima MAX ultracentrifuges, Near vertical (ultracentrifugation)
Přejít na:
Price
582 436,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
MlN-80 rotor package, near-vertical tube (9 angle), titanium.
Major applications: Rapid contamination-free isopycnic isolation of plasmid DNA. For use in Optima MAX-XP benchtop ultracentrifuges and Optima MAX ultracentrifuges.
Specifikace
  • Popis:
    MLN-80 NVT rotor package, 80000 rpm, 389000 ×g, 8×8 ml. Major applications: Rapid contamination-free isopycnic isolation of plasmid DNA. For use in Optima MAX-XP benchtop ultracentrifuges and Optima MAX ultracentrifuges
  • Určeno pro:
    Near vertical (ultracentrifugation)
  • Poloměr:
    54,2 mm
  • Cat. No.:
    521-4042
  • No. of tubes accepted:
    8×8 ml
  • Max. RPM:
    80000