Orders LinkContactUsLinkComponent
Mop head frame, small, flat, BCR® Mop 3
Mop head frame, small, flat, BCR® Mop 3
BCR® 3 Small flat mop head frame, 28×9 cm, Eloxovaný hliník
Catalog # BERKBCR.MOP3.1
Supplier:   BERKSHIRE
Mop head frame, small, flat, BCR® Mop 3
Catalog # BERKBCR.MOP3.1
Supplier:  BERKSHIRE
Mop head frame, small, flat, BCR® Mop 3
Mop head frame, small, flat, BCR® Mop 3
BCR® 3 Small flat mop head frame, 28×9 cm, Eloxovaný hliník
Price
13 300,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Berkshire’s flat mop head is light and easy to maneuver, even with wet wipers attached. The stainless steel springs or plastic eyes hold multiple mop wipers.
  • Clean, light, and durable
  • High quality anodized aluminum
  • Autoclavable (plastic eyes are not autoclavable)
  • Plastic eyes for wiper attachment
Efficient cleaning of floors, walls, and other large surfaces is critical to the overall cleanliness of the production environment. BCR® mop systems replicate the most effective way to clean a surface wiping. Berkshire’s mop systems are easy to use, highly effective, and quick.
The system is very flexible, combining a wide range of alternative mop wipers designed for particular levels of cleanliness with a range of flat mop heads.
Certifications: Designed for use in ISO class 3 to 8 environments. Cleanroom class 1 to 100000.
Specifications
  • Popis:
    BCR® 3 Small flat mop head frame
  • D׊:
    28×9 cm
  • Materiál:
    Eloxovaný hliník