Orders LinkContactUsLinkComponent
Rotor Package, Avanti JS-5.3
Rotor Package, Avanti JS-5.3
JS-5.3 swinging bucket (high performance) aluminium rotor package with microplate kit. For use in Avanti J-E and J-26 XP Series centrifuges only. Package includes rotor, buckets, tie-down knob, and microplate carrier kit (368914), Four-place rotor with anodized aluminium buckets designed for use with adapters to accommodate a wide range of tubes and bottles. Major applications: Rapid sedimentation of protein precipitates, large particles, cells, and cell debris. It can be used for binding studies and blood sample preparation
Katalogové číslo 521-4165
Dodavatel:   Beckman Coulter
Rotor Package, Avanti JS-5.3
Katalogové číslo 521-4165
Dodavatel:  Beckman Coulter
Rotor Package, Avanti JS-5.3
Rotor Package, Avanti JS-5.3
JS-5.3 swinging bucket (high performance) aluminium rotor package with microplate kit. For use in Avanti J-E and J-26 XP Series centrifuges only. Package includes rotor, buckets, tie-down knob, and microplate carrier kit (368914), Four-place rotor with anodized aluminium buckets designed for use with adapters to accommodate a wide range of tubes and bottles. Major applications: Rapid sedimentation of protein precipitates, large particles, cells, and cell debris. It can be used for binding studies and blood sample preparation
Přejít na:
Price
488 179,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
JS-5.3 Swinging-Bucket rotor package, anodized aluminum. Four-place rotor with anodized aluminum buckets designed for use with adapters to accommodate a wide range of tubes and bottles. Includes rotor, buckets, tie-down knob, and microplate carrier kit (BECM368914).
Major applications: Rapid sedimentation of protein precipitates, large particles, cells, and cell debris. It can be used for binding studies and blood sample preparation. For use in Avanti J-E and J-26 XP Series centrifuges only.
Specifikace
  • Popis:
    JS-5.3 swinging bucket (high performance) aluminium rotor package with microplate kit. For use in Avanti J-E and J-26 XP Series centrifuges only. Package includes rotor, buckets, tie-down knob, and microplate carrier kit (368914)
  • Určeno pro:
    Four-place rotor with anodized aluminium buckets designed for use with adapters to accommodate a wide range of tubes and bottles. Major applications: Rapid sedimentation of protein precipitates, large particles, cells, and cell debris. It can be used for binding studies and blood sample preparation
  • Cat. No.:
    521-4165
  • Kapacita:
    Nominal capacity - 2000 ml
  • Max. RPM:
    5300