Rotor package, Avanti J-Lite JLA-8.1000
521-4261
- Popis:J-LITE JLA-8.1000 rotor package. Includes rotor body, lid, carbon fibre canisters, labware kit (bottle rack, spatula, bottle grip and wrench 392574), and 6 polypropylene bottles with cap assemblies (3× C31597)
- Bal.:1
- Max. RPM:8000
- No. of tubes accepted:6×1000 ml/95×191 mm (3,8×7,65")