Orders LinkContactUsLinkComponent
Fixed-angle titanium rotor package/assembly, MLA-55
Fixed-angle titanium rotor package/assembly, MLA-55
Dodavatel:   Beckman Coulter
Fixed-angle titanium rotor package/assembly, MLA-55
Dodavatel:  Beckman Coulter
Certifikáty
Fixed-angle titanium rotor package/assembly, MLA-55
Fixed-angle titanium rotor package/assembly, MLA-55
2 Dostupných variant níže
Product Details & Documents
Major applications: Subcellular isolations.
Specifications for Product Family
Specifikace
  • Catalog No:
  • 521-4172
  • 521-4096
  • Kapacita:
  • 108 ml
  • 108 ml
  • No. of tubes accepted:
  • 2/4/6/8×7 ml
  • 2/4/6/8×7 ml
  • Poloměr:
  • 84,5 mm
  • 84,5 mm
  • Max. RPM:
  • 55000
  • 55000
Specifikace
Product Family Options
Informace o produktu
  • Sekvence
    Určeno pro
    Dostupnost
    Price
  • 521-4172
    MLA-55 fixed-angle titanium rotor package, includes rotor, 4 boxes of OptiSeal Tubes (BECM361623), 12 spacers (BECM361670) (6 pack. of 2), 1 OptiSeal Tube Rack (BECM361642), 1 tube extraction tool (BECM361668) and 1 spacer removal tool (BECM338765).
    MLA-55 fixed-angle titanium rotor package, includes rotor, 4 boxes of OptiSeal Tubes (BECM361623), 12 spacers (BECM361670) (6 pack. of 2), 1 OptiSeal Tube Rack (BECM361642), 1 tube extraction tool (BECM361668) and 1 spacer removal tool (BECM338765).
    01
    Optima MAX-XP benchtop ultracentrifuges and Optima MAX micro ultracentrifuges
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 634 539,00 Kč
    634 539,00 Kč
    1 * 1 KS
       634 539,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    521-4172
    Kapacita
    108 ml
    No. of tubes accepted
    2/4/6/8×7 ml
    Poloměr
    84,5 mm
    Max. RPM
    55000
  • 521-4096
    MLA-55 fixed-angle titanium rotor assembly
    MLA-55 fixed-angle titanium rotor assembly
    02
    Optima MAX-XP benchtop ultracentrifuges and Optima MAX micro ultracentrifuges
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 619 757,00 Kč
    619 757,00 Kč
    1 * 1 KS
       619 757,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    521-4096
    Kapacita
    108 ml
    No. of tubes accepted
    2/4/6/8×7 ml
    Poloměr
    84,5 mm
    Max. RPM
    55000