ContactUsLinkComponent
HPLC All-In-One box, b.safe
HPLC All-In-One box, b.safe
Katalogové číslo 590-0412
Dodavatel:  Bohlender
undefined
HPLC All-In-One box, b.safe
Katalogové číslo 590-0412
Dodavatel:  Bohlender
Specifikace
  • Popis:
    HPLC All-In-One box
  • Bal.:
    1
Specifikace
O tomto produktu
Complete set for equipping one HPLC system.
Containing b.safe Caps GL 45 for four laboratory flasks for solvent extraction as well as a b.safe waste cap with exhaust filter and a canister for safe collection of solvent waste. The scope of delivery of the b.safe caps includes PFA fittings for capillary Ext.Ø. 3,2 mm, blind fitting in PFA to close unused connections and Air Valves with service life indicator. For the collection of waste, the set includes a PE-canister S55 with 5-litre capacity, a suitable waste cap S55 and one exhaust filter L. The scope of delivery of the waste cap includes PFA Fittings for capillaries with Ext.Ø. 1,6/2,2/3,2 mm, blind fittings to close unused connections, tubing connectors for tubing Int.Ø. 5,5 to 8 mm. Consumables (air valves, exhaust filter) for a 12 months service life included, laboratory flasks GL45 have to be purchased separately.
3× b.safe caps GL 45 with 1× capillary connection, incl. 1× fitting for tubing Ext.Ø. 3,2 mm, 1 air valve with service life indicator. (Cat. No. BOHLM145-01)
1× b.safe cap GL 45 with 2× capillary connection, incl. 2× fitting for tubing Ext.Ø. 3,2 mm, 1 air valve with service life indicator. (Cat. No. BOHLM145-02)
4× b.safe air valve with service life indicator 6 months. (Cat. No. BOHLM505-01)
1× b.safe waste cap S55 with 4× capillary connection, 4× hose connector, 1× connector for exhaust filter, incl. 4× Fitting for Tubing Ext.Ø. 1,6 mm (green) / 2,2 mm (purple) / 3,2 mm (yellow), 4× blind fitting for capillary connection, 4× blind fitting hose connection, 4× hose connector for tubing Int.Ø 5,5 to 8 mm (Cat. No. BOHLM355-40)
1× b.safe hose connector for tubing Int.Ø. 9 to 10,5 mm (Cat. No. BOHLM503-16)
1× b.safe exhaust filter L, service life 12 months (Cat. No. BOHLM506-03)
1× b.safe canister S55, PE, capacity 5 litres (Cat. No. BOHLR225-05)