Powered air respirator airflow indicator TR-970
Catalog # MMMATR-970
Supplier: 3M
Specifications
- Popis:Air flow indicator
- Bal.:1
Specifications
About this item
The TR-970 Air Flow Indicator is used to confirm that the manufacturer's minimum design flow rate of the Versaflo™ TR-300 Series Powered Air Turbo is exceeded.
- Designed to enable testing at various elevations and temperatures within the operational range
- Easy to use