- Popis:Cleaning and storage kit
- Environmentally Preferable:
- Bal.:1
The Versaflo™ Cleaning and Storage Kit TR-653 is designed to assist with safely cleaning and storing the Versaflo™ Powered Air Turbo TR-600 and TR-800 Series. It comes with one inlet plug and two inlet seals, one outlet plug and two outlet seals, and one battery connector cover.
- Simplifies cleaning and storage
- Turbo plugs allow turbo unit TR-600 to be submerged or rinsed under running water
- Plugs keep the internal components of the turbo clean during storage
- Plugs and seals should be replaced when worn or damaged, or after 30 uses
For easy and safe cleaning and storage of the Versaflo™ Powered Air Turbo TR-600 and TR-800 Series, use the Versaflo™ Cleaning and Storage Kit. This cleaning and storage kit comes with a plug and two seals for both the inlet and outlet openings of TR-600/TR-800 Series turbo, and a cover for the electrical connections of the TR-600/TR-800 Series battery. The turbo plugs allow the unit to be submerged or rinsed under running water as outlined in the user instructions for easy cleaning. The plugs can also help to keep the internal components of TR-600/TR-800 Series turbo clean and free from contaminants during storage. The plug seals should be replaced after 30 uses or whenever found to be damaged or worn.