Powered air respirator visor frame assemblies, Versaflo™
Katalogové číslo MMMAM-961
Dodavatel: 3M
Specifikace
- Popis:Visor frame assemblies
- Bal.:1
Specifikace
O tomto produktu
The Versaflo™ Over-Visor Frame M-961 is a carrier frame designed to fit Versaflo™ Over-Visor M-967 and Versaflo™ radiant heat covers for Versaflo™ M-Series Helmets.
- Carrier frame to fit over-visor and radiant heat covers to M-Series helmets
- Designed specifically for use with M-Series helmets
This frame is made from aluminium.