Powered and supplied air replacement visors and lenses, Versaflo™
Katalogové číslo MMMAM927
Dodavatel: 3M
Specifikace
- Barva:Průhledná
- Popis:Replacement visors and lenses
- Bal.:1
Specifikace
O tomto produktu
The Versaflo™ coated visor M-927 is a clear, polycarbonate replacement visor designed for use with 3M™ Versaflo™ M-Series Headtops.
- Coated to provide chemical and scratch resistance
- Clear polycarbonate
- CE approved to EN 166 1:BT:3
Replace scratched and worn visors to ensure continued good vision. This replacement polycarbonate visor provides chemical and scratch resistance for 3M™ Versaflo™ M-Series Headtops.