ContactUsLinkComponent
CPC® AseptiQuik® G Series Genderless Aseptic Connectors
undefined
CPC® AseptiQuik® G Series Genderless Aseptic Connectors
  • Maximální provozní tlak:
  • 60 psi
  • 60 psi
  • 60 psi
  • 60 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • Maximální teplota:
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • Minimální teplota:
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • 1 °C (34 °F)
  • 4 °C (39 °F)
  • 1 °C (34 °F)
  • 1 °C (34 °F)
  • 1 °C (34 °F)
  • Seal material:
  • Platinim-cured silicone
  • Platinim-cured silicone
  • Platinim-cured silicone
  • Platinim-cured silicone
  • Platinum-cured silicone
  • Platinum-cured silicone
  • Platinim-cured silicone
  • Platinim-cured silicone
  • Platinim-cured silicone
  • Sterilizace:
  • Gamma and Autoclave
  • Gamma and Autoclave
  • Gamma and Autoclave
  • Gamma and Autoclave
  • Gamma and Autoclave
  • Gamma and Autoclave
  • Gamma and Autoclave
  • Gamma and Autoclave
  • Gamma and Autoclave
  • Regulatory approvals:
  • USP Class VI
  • USP Class VI
  • USP Class VI
  • USP Class VI
  • USP Class VI
  • USP Class VI
  • USP Class VI
  • USP Class VI
  • USP Class VI
Frequently Bought Together