ContactUsLinkComponent
PrimeLock® Benchtop Installation Tool, 1/4" to 1" OD tubing
Catalog # MFLX30493-25
2 month warranty
PrimeLock® Benchtop Installation Tool, 1/4" to 1" OD tubing
Catalog # MFLX30493-25
2 month warranty
Specifications
  • Popis:
    Benchtop Installation Tool, 1/4" to 1" OD tubing
  • Kompatibilní s:
    PrimeLock High-Purity PFA Flare Fittings, 1/4 to 1" OD Tubing
  • Bal.:
    1
  • Záruka:
    2 months
Specifications
About this item
Easily install your High-Purity PFA Fittings.
  • Optimize performance of your PFA PrimeLock® Fittings
Handheld and benchtop installation tools help you to quickly and easily install the PrimeLock insert correctly every time. Benchtop tool is the quickest and easiest method of installing the insert when space constraints are not an issue. The compact, handheld tool enables you to install the insert in remote locations. Installation tools require mandrel(s) and clamp insert(s) for operation.
  • Popis:
    Benchtop Installation Tool, 1/4" to 1" OD tubing
  • Environmentally Preferable:
  • Kompatibilní s:
    PrimeLock High-Purity PFA Flare Fittings, 1/4 to 1" OD Tubing
  • Bal.:
    1
  • Záruka:
    2 months