Masterflex® Close-Coupled Industrial Gear Pump Heads, Avantor®
Rugged corrosion-resistant bronze construction provides long service life.
- Easy assembly to NEMA and IEC motor frames
These gear pump heads are suitable for all liquids that are compatible with bronze—most common liquids are water, oil, and mild chemicals (pH from 4 to 11).
Choose a gear pump head to mount to your own motor (adapters for NEMA or IEC motors are sold separately below) or select a complete system with the pump head conveniently mounted to a standard (NEMA 56C) TEFC motor.
- Environmentally Preferable:
- Materiál trupu:
- Bronze
- Bronze
- Bronze
- Bronze
- Bronze
- Gear material:
- Bronze
- Bronze
- Bronze
- Bronze
- Bronze
- Inlet connection:
- 0,375 NPT(F)
- 0,25 NPT(F)
- 0,375 NPT(F)
- 0,25 NPT(F)
- 0,25 NPT(F)
- Maximální tlak:
- 10,3 bar (150 psi)
- 10,3 bar (150 psi)
- 10,3 bar (150 psi)
- Maximální teplota:
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- Minimální teplota:
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- Outlet connection:
- 0,375 NPT(F)
- 0,25 NPT(F)
- 0,375 NPT(F)
- 0,25 NPT(F)
- 0,25 NPT(F)
- Relief valve:
- No
- No
- Yes
- No
- Yes
- Suction lift, wet:
- 6096 mm of H₂O (240" H₂O)
- 6096 mm of H₂O (240" H₂O)
- 6096 mm of H₂O (240" H₂O)
- 6096 mm of H₂O (240" H₂O)
- 6096 mm of H₂O (240" H₂O)
- Máčené materiály:
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Záruka:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Frequently Bought Together